Tisk…

Triethylene glycol monobenzyl ether 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol, 2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethanol

OR3392-25G
APOSOR3392-25GEA 1580 CZK
APOSOR3392-25G
Triethylene glycol monobenzyl ether 98%
Triethylene glycol monobenzyl ether
Vzorec: C₁₃H₂₀O₄
MW: 240,30 g/mol
Bod varu: 126 °C/0,075mmHg
Hustota: 1,09 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093556
CAS číslo: 55489-58-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR