2-Bromo-6-methoxyaniline 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR3368-1G
OR3368-5G
OR3368-25G
APOSOR3368-1GEA
1660
CZK
APOSOR3368-1G
APOSOR3368-5G
APOSOR3368-25G
2-Bromo-6-methoxyaniline 97%
2-Bromo-6-methoxyaniline
Vzorec:
C₇H₈BrNO MW: 202,0500 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08741448 CAS číslo: 5473-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...