Tisk…

4-Bromo-2-hydrazinopyridine

Dodavatel: Apollo Scientific
Varování

OR33676-1G OR33676-5G
APOSOR33676-1GEA 3440 CZK
APOSOR33676-1G APOSOR33676-5G
4-Bromo-2-hydrazinopyridine
4-Bromo-2-hydrazinopyridine
Vzorec: C₅H₆BrN₃
Storage Temperature: Freezer
MDL Number: MFCD00234091
CAS číslo: 1019918-39-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR