Benzoin methyl ether 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone
OR33657-100G
OR33657-25G
OR33657-500G
APOSOR33657-100GEA
4960
CZK
APOSOR33657-100G
APOSOR33657-25G
APOSOR33657-500G
Benzoin methyl ether 96%
Benzoin methyl ether
Vzorec:
C₁₅H₁₄O₂ MW: 226,27 g/mol Bod tání: 47…50 °C Hustota: 1,128 g/cm³ (25 °C) Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008492 CAS číslo: 3524-62-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...