Tisk…

Tetrabutylammonium chloride 98%

Dodavatel: Apollo Scientific
Varování

OR33653-1KG OR33653-250G
APOSOR33653-1KGEA 16530 CZK
APOSOR33653-1KG APOSOR33653-250G
Tetrabutylammonium chloride 98%
Tetrabutylammonium chloride
Vzorec: C₁₆H₃₆ClN
Storage Temperature: Ambient
MDL Number: MFCD00011635
CAS číslo: 1112-67-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR