Tisk…

N-[(E)-(2-Chloro-5-nitrophenyl)methylidene]methanamine 95%

Dodavatel: Apollo Scientific

OR33639-1G OR33639-5G
APOSOR33639-1GEA 5130 CZK
APOSOR33639-1G APOSOR33639-5G
N-[(E)-(2-Chloro-5-nitrophenyl)methylidene]methanamine 95%
N-[(E)-(2-Chloro-5-nitrophenyl)methylidene]methanamine
Vzorec: C₈H₇ClN₂O₂
MW: 198,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07681942
CAS číslo: 871909-85-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR