2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione 95%
Dodavatel: Apollo Scientific
OR33626-1G
OR33626-25G
OR33626-5G
OR33626-10G
APOSOR33626-1GEA
5130
CZK
APOSOR33626-1G
APOSOR33626-25G
APOSOR33626-5G
APOSOR33626-10G
2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione 95%
2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione
Vzorec:
C₁₃H₁₃NO₃ MW: 231,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797717 CAS číslo: 3197-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...