Tisk…

Methyl-1-cyanocyclopropanecarboxylate 95%

Dodavatel: Apollo Scientific

OR33622-1G OR33622-5G
APOSOR33622-1GEA 9560 CZK
APOSOR33622-1G APOSOR33622-5G
Methyl-1-cyanocyclopropanecarboxylate 95%
Methyl-1-cyanocyclopropanecarboxylate
Vzorec: C₆H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD09033250
CAS číslo: 6914-73-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR