Tisk…

3-Methyl-1H-pyrazolo[3,4-b]pyridine 95%

Dodavatel: Apollo Scientific

OR33616-5G
APOSOR33616-5GEA 3060 CZK
APOSOR33616-5G
3-Methyl-1H-pyrazolo[3,4-b]pyridine 95%
3-Methyl-1H-pyrazolo[3,4-b]pyridine
Vzorec: C7H7N3
MW: 133,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607943
CAS číslo: 116834-96-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR