Tisk…

[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Isopropylisoxazol-5-amine

OR33606-1G OR33606-5G
APOSOR33606-1GEA 2530 CZK
APOSOR33606-1G APOSOR33606-5G
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine 97%
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine
Vzorec: C₆H₁₀N₂O
MW: 126,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06335179
CAS číslo: 88786-11-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR