Tisk…

1-{1H-Pyrrolo[2,3-b]pyridin-3-ylmethyl}piperidine 97%

Dodavatel: Apollo Scientific

OR33602-1G OR33602-5G
APOSOR33602-1GEA 9560 CZK
APOSOR33602-1G APOSOR33602-5G
1-{1H-Pyrrolo[2,3-b]pyridin-3-ylmethyl}piperidine 97%
1-{1H-Pyrrolo[2,3-b]pyridin-3-ylmethyl}piperidine
Vzorec: C₁₃H₁₇N₃
MW: 215,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22126110
CAS číslo: 23616-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR