4-tert-Butyl-N-methyl-1,3-thiazol-2-aminehydrobromide 95%
Dodavatel: Apollo Scientific
OR33592-1G
OR33592-5G
APOSOR33592-1GEA
7670
CZK
APOSOR33592-1G
APOSOR33592-5G
4-tert-Butyl-N-methyl-1,3-thiazol-2-aminehydrobromide 95%
4-tert-Butyl-N-methyl-1,3-thiazol-2-aminehydrobromide
Vzorec:
C₈H₁₅BrN₂S Storage Temperature: Ambient |
MDL Number:
MFCD18803537 CAS číslo: 1280787-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...