2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(3,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile
OR33579-1G
OR33579-5G
APOSOR33579-1GEA
7670
CZK
APOSOR33579-1G
APOSOR33579-5G
2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile 95%
2-(3,4-Dichlorophenyl)-3-hydroxyacrylonitrile
Vzorec:
C₉H₅Cl₂NO MW: 214,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12025879 CAS číslo: 77186-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...