4-Iodo-1H-pyrrole-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR33575-1G
OR33575-5G
APOSOR33575-1GEA
15310
CZK
APOSOR33575-1G
APOSOR33575-5G
4-Iodo-1H-pyrrole-2-carboxylic acid 95%
4-Iodo-1H-pyrrole-2-carboxylic acid
Vzorec:
C₅H₄INO₂ Storage Temperature: Ambient |
MDL Number:
MFCD10758077 CAS číslo: 252861-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...