N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 97%
Dodavatel: Apollo Scientific
Synonyms:
7-Azagramine, 3-(Dimethylaminomethyl)-1H-pyrrolo[2,3-b]pyridine,7-Azagramine,Dimethyl-(1H-pyrrolo[2,3-b]-3-pyridinylmethyl)amine
OR33563-1G
OR33563-5G
APOSOR33563-1GEA
2610
CZK
APOSOR33563-1G
APOSOR33563-5G
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 97%
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine
Vzorec:
C₁₀H₁₃N₃ MW: 175,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01580034 CAS číslo: 5654-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...