Tisk…

8-Methyl-1-thia-4-azaspiro[4.5]decane 95%

Dodavatel: Apollo Scientific

OR33549-1G OR33549-5G
APOSOR33549-1GEA 7670 CZK
APOSOR33549-1G APOSOR33549-5G
8-Methyl-1-thia-4-azaspiro[4.5]decane 95%
8-Methyl-1-thia-4-azaspiro[4.5]decane
Vzorec: C₉H₁₇NS
Storage Temperature: Ambient
MDL Number: MFCD16140310
CAS číslo: 31404-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR