4-Isopropylbenzenecarbothioamide 95%
Dodavatel: Apollo Scientific
OR33532-1G
OR33532-5G
APOSOR33532-1GEA
7670
CZK
APOSOR33532-1G
APOSOR33532-5G
4-Isopropylbenzenecarbothioamide 95%
4-Isopropylbenzenecarbothioamide
Vzorec:
C₁₀H₁₃NS Storage Temperature: Ambient |
MDL Number:
MFCD05264094 CAS číslo: 53515-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...