Tisk…

3-Bromo-1-methyl-1H-indole-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR33530-1G
APOSOR33530-1GEA 9560 CZK
APOSOR33530-1G
3-Bromo-1-methyl-1H-indole-2-carboxylic acid 95%
3-Bromo-1-methyl-1H-indole-2-carboxylic acid
Vzorec: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11099656
CAS číslo: 499983-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR