3-(1-Benzofuran-2-yl)-3-oxopropanenitrile 95%
Dodavatel: Apollo Scientific
OR33529-1G
OR33529-5G
APOSOR33529-1GEA
7670
CZK
APOSOR33529-1G
APOSOR33529-5G
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile 95%
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile
Vzorec:
C₁₁H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00047286 CAS číslo: 5149-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...