3-(1H-Indol-3-yl)-3-oxopropanenitrile 95%
Dodavatel: Apollo Scientific
Synonyms:
3-(Cyanoacetyl)indole
OR33521-25G
OR33521-5G
APOSOR33521-25GEA
37030
CZK
APOSOR33521-25G
APOSOR33521-5G
3-(1H-Indol-3-yl)-3-oxopropanenitrile 95%
3-(1H-Indol-3-yl)-3-oxopropanenitrile
Vzorec:
C₁₁H₈N₂O MW: 184,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04610443 CAS číslo: 20356-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...