Tisk…

2-(4-Methoxyphenyl)-2-oxoethyl acetate 95%

Dodavatel: Apollo Scientific

OR33512-1G OR33512-5G
APOSOR33512-1GEA 5130 CZK
APOSOR33512-1G APOSOR33512-5G
2-(4-Methoxyphenyl)-2-oxoethyl acetate 95%
2-(4-Methoxyphenyl)-2-oxoethyl acetate
Vzorec: C₁₁H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00540316
CAS číslo: 58518-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR