Tisk…

tert-Butyl-2-(2-cyanophenoxy)acetate 95%

Dodavatel: Apollo Scientific

OR33509-1G OR33509-5G
APOSOR33509-1GEA 5130 CZK
APOSOR33509-1G APOSOR33509-5G
tert-Butyl-2-(2-cyanophenoxy)acetate 95%
tert-Butyl-2-(2-cyanophenoxy)acetate
Vzorec: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972227
CAS číslo: 609804-44-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR