Tisk…

8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one 95%

Dodavatel: Apollo Scientific

OR33505-1G OR33505-5G
APOSOR33505-1GEA 7670 CZK
APOSOR33505-1G APOSOR33505-5G
8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one 95%
8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one
Vzorec: C₁₄H₁₈N₂OS
Storage Temperature: Ambient
MDL Number: MFCD16140266
CAS číslo: 32533-11-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR