1-Thia-4-azaspiro[4.6]undecan-3-one 95%
Dodavatel: Apollo Scientific
OR33500-1G
OR33500-5G
APOSOR33500-1GEA
7670
CZK
APOSOR33500-1G
APOSOR33500-5G
1-Thia-4-azaspiro[4.6]undecan-3-one 95%
1-Thia-4-azaspiro[4.6]undecan-3-one
Vzorec:
C₉H₁₅NOS Storage Temperature: Ambient |
MDL Number:
MFCD00101409 CAS číslo: 1221791-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...