2-Chlorophenylboronic acid 97+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
OR3349-100G
OR3349-25G
OR3349-5G
APOSOR3349-100GEA
4070
CZK
APOSOR3349-100G
APOSOR3349-25G
APOSOR3349-5G
2-Chlorophenylboronic acid 97+%
2-Chlorophenylboronic acid
Vzorec:
C₆H₆BClO₂ MW: 156.38 Bod tání: 92…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674012 CAS číslo: 3900-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...