3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile 95%
Dodavatel: Apollo Scientific
OR33478-1G
OR33478-5G
APOSOR33478-1GEA
9560
CZK
APOSOR33478-1G
APOSOR33478-5G
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile 95%
3-[4-(4-Methylpiperazino)phenyl]-3-oxopropanenitrile
Vzorec:
C₁₄H₁₇N₃O MW: 243,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12025842 CAS číslo: 1135282-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...