3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile 95%
Dodavatel: Apollo Scientific
OR33472-1G
OR33472-5G
APOSOR33472-1GEA
6950
CZK
APOSOR33472-1G
APOSOR33472-5G
3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile 95%
3-Oxo-3-(1H-pyrrol-2-yl)propanenitrile
Vzorec:
C₇H₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD07369265 CAS číslo: 90908-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...