Tisk…

3-Bromo-5-methylpyridin-2-ol 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromo-2-hydroxy-5-methylpyridine

OR3347-1G OR3347-5G
APOSOR3347-1GEA 622 CZK
APOSOR3347-1G APOSOR3347-5G
3-Bromo-5-methylpyridin-2-ol 98%
3-Bromo-5-methylpyridin-2-ol
Vzorec: C6H6BrNO
MW: 188,02 g/mol
Bod tání: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03791312
CAS číslo: 17282-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR