Tisk…

6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine 95%

Dodavatel: Apollo Scientific

OR33469-1G OR33469-5G
APOSOR33469-1GEA 9560 CZK
APOSOR33469-1G APOSOR33469-5G
6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine 95%
6-Chloro-N-methyl-2-phenyl-4-pyrimidinamine
Vzorec: C₁₁H₁₀ClN₃
MW: 219,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00697490
CAS číslo: 1017782-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR