Tisk…

3-(4-Chlorophenyl)-1H-pyrazol-4-amine 95%

Dodavatel: Apollo Scientific

OR33467-1G
APOSOR33467-1GEA 9560 CZK
APOSOR33467-1G
3-(4-Chlorophenyl)-1H-pyrazol-4-amine 95%
3-(4-Chlorophenyl)-1H-pyrazol-4-amine
Vzorec: C₉H₈ClN₃
MW: 193,632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10758055
CAS číslo: 91857-91-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR