Tisk…

7-Acetyl-2-methylbenzoxazole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(2-Methyl-1,3-benzoxazol-7-yl)ethan-1-one

OR33455-1G OR33455-5G
APOSOR33455-1GEA 7670 CZK
APOSOR33455-1G APOSOR33455-5G
7-Acetyl-2-methylbenzoxazole 95%
7-Acetyl-2-methylbenzoxazole
Vzorec: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD09152710
CAS číslo: 952182-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR