Tisk…

Methyl-2-chloro-4-(1H-imidazol-1-yl)benzenecarboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-chloro-4-(1H-imidazol-1-yl)benzoate

OR33452-1G OR33452-5G
APOSOR33452-1GEA 7670 CZK
APOSOR33452-1G APOSOR33452-5G
Methyl-2-chloro-4-(1H-imidazol-1-yl)benzenecarboxylate 95%
Methyl-2-chloro-4-(1H-imidazol-1-yl)benzenecarboxylate
Vzorec: C₁₁H₉ClN₂O₂
MW: 236,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09152705
CAS číslo: 952182-93-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR