4-(2,6-Dimethylmorpholino)aniline 95%
Dodavatel: Apollo Scientific
OR33419-1G
OR33419-5G
APOSOR33419-1GEA
7670
CZK
APOSOR33419-1G
APOSOR33419-5G
4-(2,6-Dimethylmorpholino)aniline 95%
4-(2,6-Dimethylmorpholino)aniline
Vzorec:
C₁₂H₁₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00051574 CAS číslo: 218930-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...