2,6-Dimethyl-4-(4-nitrophenyl)morpholine 95%
Dodavatel: Apollo Scientific
OR33418-1G
OR33418-25G
OR33418-5G
APOSOR33418-1GEA
5130
CZK
APOSOR33418-1G
APOSOR33418-25G
APOSOR33418-5G
2,6-Dimethyl-4-(4-nitrophenyl)morpholine 95%
2,6-Dimethyl-4-(4-nitrophenyl)morpholine
Vzorec:
C₁₂H₁₆N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00105780 CAS číslo: 29842-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...