Tisk…

Methyl N-(4-methoxyphenyl)carbamate 95%

Dodavatel: Apollo Scientific

OR33413-1G OR33413-5G
APOSOR33413-1GEA 7670 CZK
APOSOR33413-1G APOSOR33413-5G
Methyl N-(4-methoxyphenyl)carbamate 95%
Methyl N-(4-methoxyphenyl)carbamate
Vzorec: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00025852
CAS číslo: 14803-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR