1-(4-Benzylpiperazino)-1-propanone 95%
Dodavatel: Apollo Scientific
OR33410-1G
OR33410-5G
APOSOR33410-1GEA
5130
CZK
APOSOR33410-1G
APOSOR33410-5G
1-(4-Benzylpiperazino)-1-propanone 95%
1-(4-Benzylpiperazino)-1-propanone
Vzorec:
C₁₄H₂₀N₂O MW: 232,33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03381901 CAS číslo: 314728-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...