1-[4-(2-Propynyloxy)phenyl]-1-ethanone 95%
Dodavatel: Apollo Scientific
OR33399-1G
OR33399-5G
APOSOR33399-1GEA
7670
CZK
APOSOR33399-1G
APOSOR33399-5G
1-[4-(2-Propynyloxy)phenyl]-1-ethanone 95%
1-[4-(2-Propynyloxy)phenyl]-1-ethanone
Vzorec:
C₁₁H₁₀O₂ MW: 174,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001308 CAS číslo: 34264-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...