2-(2-Propynyloxy)benzenecarbaldehyde oxime 95%
Dodavatel: Apollo Scientific
OR33397-1G
OR33397-5G
APOSOR33397-1GEA
9560
CZK
APOSOR33397-1G
APOSOR33397-5G
2-(2-Propynyloxy)benzenecarbaldehyde oxime 95%
2-(2-Propynyloxy)benzenecarbaldehyde oxime
Vzorec:
C₁₀H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03001307 CAS číslo: 55241-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...