Tisk…

4-Acetyl-2-(trichloroacetyl)pyrrole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(4-Acetyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

OR33396-1G OR33396-5G
APOSOR33396-1GEA 15310 CZK
APOSOR33396-1G APOSOR33396-5G
4-Acetyl-2-(trichloroacetyl)pyrrole 95%
4-Acetyl-2-(trichloroacetyl)pyrrole
Vzorec: C₈H₆Cl₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD03001304
CAS číslo: 72652-34-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR