Tisk…

[2-(2-Propynyloxy)phenyl]methanol 95%

Dodavatel: Apollo Scientific

OR33395-1G OR33395-5G
APOSOR33395-1GEA 9560 CZK
APOSOR33395-1G APOSOR33395-5G
[2-(2-Propynyloxy)phenyl]methanol 95%
[2-(2-Propynyloxy)phenyl]methanol
Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001302
CAS číslo: 130441-68-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR