Tisk…

3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide 95%

Dodavatel: Apollo Scientific

OR33366-1G OR33366-25G OR33366-5G
APOSOR33366-1GEA 5130 CZK
APOSOR33366-1G APOSOR33366-25G APOSOR33366-5G
3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide 95%
3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide
Vzorec: C₁₃H₁₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00793443
CAS číslo: 454473-71-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR