3-{4-(Methylsulphonyl)benzene}-1-propanoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR3336-1G
OR3336-5G
APOSOR3336-1GEA
1120
CZK
APOSOR3336-1G
APOSOR3336-5G
3-{4-(Methylsulphonyl)benzene}-1-propanoic acid 98%
3-{4-(Methylsulphonyl)benzene}-1-propanoic acid
Vzorec:
C₁₀H₁₂O₄S Bod tání: 173…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319236 CAS číslo: 387350-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...