Tisk…

2-Chloro-2'-methoxyacetanilide 95%

Dodavatel: Apollo Scientific

OR33345-25G OR33345-5G
APOSOR33345-25GEA 23110 CZK
APOSOR33345-25G APOSOR33345-5G
2-Chloro-2'-methoxyacetanilide 95%
2-Chloro-2'-methoxyacetanilide
Vzorec: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00095394
CAS číslo: 55860-22-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR