Tisk…

1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Dodavatel: Apollo Scientific

OR33338-1G OR33338-5G
APOSOR33338-1GEA 4630 CZK
APOSOR33338-1G APOSOR33338-5G
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(2,4-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Vzorec: C₁₃H₉Cl₂NO₃
MW: 298,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140302
CAS číslo: 338977-51-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR