4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%
Dodavatel: Apollo Scientific
OR33331-1G
OR33331-5G
APOSOR33331-1GEA
5130
CZK
APOSOR33331-1G
APOSOR33331-5G
4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%
4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Vzorec:
C₁₁H₆BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00139239 CAS číslo: 7721-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...