3-Phenoxy-2-butanone 95%
Dodavatel: Apollo Scientific
OR33326-1G
OR33326-5G
APOSOR33326-1GEA
7670
CZK
APOSOR33326-1G
APOSOR33326-5G
3-Phenoxy-2-butanone 95%
3-Phenoxy-2-butanone
Vzorec:
C₁₀H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00243672 CAS číslo: 6437-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...