Tisk…

2-(3-Iodophenoxy)pyrimidine 95%

Dodavatel: Apollo Scientific

OR33316-1G OR33316-25G OR33316-5G
APOSOR33316-1GEA 5130 CZK
APOSOR33316-1G APOSOR33316-25G APOSOR33316-5G
2-(3-Iodophenoxy)pyrimidine 95%
2-(3-Iodophenoxy)pyrimidine
Vzorec: C₁₀H₇IN₂O
Storage Temperature: Ambient
MDL Number: MFCD08056676
CAS číslo: 886362-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR