1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane 97%
Dodavatel: Apollo Scientific
OR33311-1G
OR33311-5G
APOSOR33311-1GEA
7670
CZK
APOSOR33311-1G
APOSOR33311-5G
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane 97%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane
Vzorec:
C₁₀H₁₅ClN₂S Storage Temperature: Ambient |
MDL Number:
MFCD06659601 CAS číslo: 885950-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...