Tisk…

5-Methyl-3-(3-thienyl)-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR33310-1G OR33310-5G
APOSOR33310-1GEA 9560 CZK
APOSOR33310-1G APOSOR33310-5G
5-Methyl-3-(3-thienyl)-1H-pyrazole 95%
5-Methyl-3-(3-thienyl)-1H-pyrazole
Vzorec: C₈H₈N₂S
MW: 164,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659599
CAS číslo: 3196-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR