Tisk…

5-Methoxy-2-(1H-pyrrol-1-yl)aniline 95%

Dodavatel: Apollo Scientific

OR33286-1G OR33286-5G
APOSOR33286-1GEA 7670 CZK
APOSOR33286-1G APOSOR33286-5G
5-Methoxy-2-(1H-pyrrol-1-yl)aniline 95%
5-Methoxy-2-(1H-pyrrol-1-yl)aniline
Vzorec: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00047063
CAS číslo: 59194-26-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR